Search
Search

TRM82| Gene Symbol: TRM82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100193.4 | NP_563675.1 | 6 - 7 | 799 | 71 | |
| GenBank mRNA | AF370228.1 | - | 6 - 7 | 798 | 71 | |
| AY059120.1 | - | 6 - 7 | 686 | 71 | ||
Set Membership: |
|