Search
Search

CRTISO| Gene Symbol: CRTISO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331668.1 | NP_001322208.1 | 10 - 11 | 2094 | 84 | |
| NM_001331669.1 | NP_001322209.1 | 11 - 12 | 1933 | 84 | ||
| NM_001331670.1 | NP_001322210.1 | 11 - 12 | 1913 | 84 | ||
| NM_100559.4 | NP_172167.2 | 11 - 12 | 2172 | 84 | ||
Set Membership: |
|