Search
Search

SLP1| Gene Symbol: SLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123768.1 | NP_001117240.1 | 7 - 8 | 1094 | 79 | |
| NM_001160844.1 | NP_001154316.1 | 8 - 9 | 1111 | 79 | ||
| NM_100574.5 | NP_172182.2 | 7 - 8 | 1160 | 79 | ||
| GenBank mRNA | AY136428.1 | - | 7 - 8 | 1019 | 79 | |
| BT008476.1 | - | 7 - 8 | 1009 | 79 | ||
Set Membership: |
|