Search
Search

RAD51C| Gene Symbol: RAD51C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202820.1 | NP_001189749.1 | 4 - 5 | 589 | 66 | |
| NM_130091.2 | NP_566040.1 | 4 - 5 | 517 | 66 | ||
| GenBank mRNA | AB062456.1 | - | 4 - 5 | 517 | 66 | |
Set Membership: |
|