Search
Search

PMM| Gene Symbol: PMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130142.4 | NP_182103.1 | 9 - 10 | 886 | 95 | |
| GenBank mRNA | AY050806.1 | - | 9 - 10 | 773 | 95 | |
| AY088930.1 | - | 9 - 10 | 782 | 95 | ||
| AY113964.1 | - | 9 - 10 | 685 | 95 | ||
| DQ442991.1 | - | 9 - 10 | 685 | 95 | ||
Set Membership: |
|