Search
Search

GPX4| Gene Symbol: GPX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337302.1 | NP_001324284.1 | 3 - 4 | 547 | 94 | |
| NM_130382.2 | NP_566128.1 | 4 - 5 | 447 | 94 | ||
| GenBank mRNA | AK118448.1 | - | 4 - 5 | 368 | 94 | |
| AY088692.1 | - | 4 - 5 | 391 | 94 | ||
| BT003735.1 | - | 4 - 5 | 317 | 94 | ||
Set Membership: |
|