Search
Search

CRB| Gene Symbol: CRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331830.1 | NP_001322308.1 | 2 - 3 | 805 | 73 | |
| NM_100804.4 | NP_172405.1 | 2 - 3 | 465 | 73 | ||
| GenBank mRNA | AF325043.2 | - | 2 - 3 | 237 | 73 | |
| AF428282.1 | - | 2 - 3 | 250 | 73 | ||
| AY035050.1 | - | 2 - 3 | 307 | 73 | ||
| AY070022.1 | - | 2 - 3 | 233 | 73 | ||
Set Membership: |
|