Search
Search

ECA3| Gene Symbol: ECA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100887.3 | NP_563860.1 | 25 - 26 | 2409 | 148 | |
| GenBank mRNA | AK229199.1 | - | 25 - 26 | 2409 | 148 | |
| EU082212.1 | - | 25 - 26 | 2220 | 148 | ||
Set Membership: |
|