Search
Search

SNL6| Gene Symbol: SNL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331906.1 | NP_001321764.1 | 12 - 13 | 2346 | 89 | |
| NM_001331907.1 | NP_001321765.1 | 11 - 12 | 2133 | 89 | ||
| NM_100919.5 | NP_172515.2 | 12 - 13 | 2346 | 89 | ||
Set Membership: |
|