Search
Search

ARFA1F| Gene Symbol: ARFA1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331919.1 | NP_001321560.1 | 2 - 3 | 481 | 100 | |
| NM_100939.4 | NP_172533.2 | 2 - 3 | 350 | 100 | ||
| GenBank mRNA | AK226308.1 | - | 2 - 3 | 251 | 100 | |
| BT014776.1 | - | 2 - 3 | 200 | 100 | ||
| BT015004.1 | - | 1 - 2 | 152 | 100 | ||
Set Membership: |
|