Search
Search

AT1G10900| Gene Symbol: AT1G10900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331939.1 | NP_001322318.1 | 6 - 7 | 2498 | 130 | |
| NM_001331940.1 | NP_001322319.1 | 7 - 8 | 2015 | 130 | ||
| NM_100965.5 | NP_172559.2 | 7 - 8 | 1990 | 130 | ||
| GenBank mRNA | AJ009782.1 | - | 7 - 8 | 1849 | 130 | |
| AY062718.1 | - | 7 - 8 | 1881 | 130 | ||
| BT010342.1 | - | 5 - 6 | 1664 | 130 | ||
Set Membership: |
|