Search
Search

HA3| Gene Symbol: HA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203630.1 | NP_001190559.1 | 8 - 9 | 1583 | 66 | |
| NM_001345258.1 | NP_001331203.1 | 4 - 5 | 1362 | 66 | ||
| NM_001345259.1 | NP_001331204.1 | 8 - 9 | 1637 | 66 | ||
| NM_125118.3 | NP_200545.1 | 8 - 9 | 1699 | 66 | ||
| GenBank mRNA | AY072153.1 | - | 8 - 9 | 1579 | 66 | |
Set Membership: |
|