Search
Search

BIO1| Gene Symbol: BIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125140.4 | NP_200567.2 | 1 - 2 | 774 | 67 | |
| GenBank mRNA | EU089963.1 | - | 1 - 2 | 769 | 67 | |
| HQ857557.1 | - | 1 - 2 | 710 | 67 | ||
Set Membership: |
|