Search
Search

CER3| Gene Symbol: CER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125164.3 | NP_200588.2 | 5 - 6 | 1276 | 63 | |
| GenBank mRNA | AB099512.1 | - | 5 - 6 | 1248 | 63 | |
| AB116548.1 | - | 5 - 6 | 957 | 63 | ||
| AY131334.1 | - | 5 - 6 | 1247 | 63 | ||
Set Membership: |
|