Search
Search

AT5G59610| Gene Symbol: AT5G59610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161316.1 | NP_001154788.1 | 4 - 5 | 612 | 107 | |
| NM_001345360.1 | NP_001318838.1 | 4 - 5 | 2597 | 107 | ||
| NM_001345361.1 | NP_001330548.1 | 4 - 5 | 2881 | 107 | ||
| NM_001345362.1 | NP_001330549.1 | 5 - 6 | 2601 | 107 | ||
| NM_125353.3 | NP_200769.1 | 3 - 4 | 2848 | 107 | ||
| GenBank mRNA | BT030000.1 | - | 3 - 4 | 583 | 107 | |
Set Membership: |
|