Search
Search

VIP2| Gene Symbol: VIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345367.1 | NP_001329954.1 | 5 - 6 | 960 | 78 | |
| NM_001345368.1 | NP_001329955.1 | 4 - 5 | 1431 | 78 | ||
| NM_125363.3 | NP_568912.2 | 4 - 5 | 1460 | 78 | ||
Set Membership: |
|