Search
Search

AT5G61140| Gene Symbol: AT5G61140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203655.2 | NP_001190584.1 | 12 - 13 | 2082 | 81 | |
| NM_125507.2 | NP_200922.2 | 12 - 13 | 1989 | 81 | ||
Set Membership: |
|