Search
Search

ATG3| Gene Symbol: ATG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345476.1 | NP_001331033.1 | 4 - 5 | 676 | 72 | |
| NM_125543.5 | NP_568934.1 | 4 - 5 | 676 | 72 | ||
| GenBank mRNA | AB073170.1 | - | 4 - 5 | 600 | 72 | |
| BT024713.1 | - | 4 - 5 | 511 | 72 | ||
Set Membership: |
|