Search
Search

ATG3| Gene Symbol: ATG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345476.1 | NP_001331033.1 | 7 - 8 | 1135 | 101 | |
| NM_125543.5 | NP_568934.1 | 8 - 9 | 1007 | 101 | ||
| GenBank mRNA | AB073170.1 | - | 8 - 9 | 931 | 101 | |
| BT024713.1 | - | 8 - 9 | 842 | 101 | ||
Set Membership: |
|