Search
Search

PFK4| Gene Symbol: PFK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037043.1 | NP_001032120.1 | 2 - 3 | 359 | 96 | |
| NM_125551.4 | NP_200966.2 | 2 - 3 | 409 | 96 | ||
| GenBank mRNA | AY099694.1 | - | 2 - 3 | 357 | 96 | |
| AY128873.1 | - | 2 - 3 | 300 | 96 | ||
Set Membership: |
|