Search
Search

GUT1| Gene Symbol: GUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125578.3 | NP_568941.1 | 3 - 4 | 1175 | 134 | |
| GenBank mRNA | AY054180.1 | - | 3 - 4 | 966 | 134 | |
| AY066039.1 | - | 3 - 4 | 735 | 134 | ||
| KJ138631.1 | - | 3 - 4 | 735 | 134 | ||
Set Membership: |
|