Search
Search

ERL1| Gene Symbol: ERL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203666.1 | NP_001190595.1 | 12 - 13 | 1246 | 70 | |
| NM_125617.3 | NP_201029.1 | 12 - 13 | 1239 | 70 | ||
| GenBank mRNA | FJ708810.1 | - | 12 - 13 | 942 | 70 | |
Set Membership: |
|