Search
Search

ALDH12A1| Gene Symbol: ALDH12A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125647.4 | NP_568955.1 | 12 - 13 | 1284 | 77 | |
| GenBank mRNA | AY065072.1 | - | 12 - 13 | 1181 | 77 | |
| BT000746.1 | - | 12 - 13 | 1214 | 77 | ||
| BT010391.1 | - | 12 - 13 | 1131 | 77 | ||
Set Membership: |
|