Search
Search

GSTF14| Gene Symbol: GSTF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103873.5 | NP_175408.1 | 2 - 3 | 389 | 65 | |
| GenBank mRNA | AK176306.1 | - | 2 - 3 | 265 | 65 | |
| BT010408.1 | - | 2 - 3 | 205 | 65 | ||
Set Membership: |
|