Search
Search

TOR| Gene Symbol: TOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123987.1 | NP_001117459.1 | 14 - 15 | 2568 | 114 | |
| NM_103891.4 | NP_175425.2 | 14 - 15 | 2568 | 114 | ||
| GenBank mRNA | AF178967.1 | - | 14 - 15 | 2423 | 114 | |
Set Membership: |
|