Search
Search

POLGAMMA2| Gene Symbol: POLGAMMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333457.1 | NP_001321058.1 | 10 - 11 | 3138 | 108 | |
| NM_103965.5 | NP_175498.2 | 10 - 11 | 3090 | 108 | ||
Set Membership: |
|