Search
Search

NRPB4| Gene Symbol: NRPB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121029.3 | NP_196554.1 | 2 - 3 | 246 | 74 | |
| GenBank mRNA | AF016511.1 | - | 2 - 3 | 193 | 74 | |
| AK317113.1 | - | 2 - 3 | 246 | 74 | ||
| AY086086.1 | - | 2 - 3 | 246 | 74 | ||
| BT025500.1 | - | 2 - 3 | 153 | 74 | ||
Set Membership: |
|