Search
Search

SULTR2;1| Gene Symbol: SULTR2;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121056.3 | NP_196580.1 | 5 - 6 | 1455 | 58 | |
| GenBank mRNA | AY062546.1 | - | 5 - 6 | 1328 | 58 | |
| AY093335.1 | - | 5 - 6 | 1247 | 58 | ||
Set Membership: |
|