Search
Search

GRF6| Gene Symbol: GRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036791.2 | NP_001031868.1 | 1 - 2 | 568 | 77 | |
| NM_001203346.1 | NP_001190275.1 | 1 - 2 | 568 | 77 | ||
| NM_001203347.1 | NP_001190276.1 | 1 - 2 | 568 | 77 | ||
| NM_121083.4 | NP_568229.1 | 1 - 2 | 758 | 77 | ||
| GenBank mRNA | AY052293.1 | - | 1 - 2 | 538 | 77 | |
| AY061918.1 | - | 1 - 2 | 482 | 77 | ||
| U02565.1 | - | 1 - 2 | 516 | 77 | ||
| U68545.1 | - | 1 - 2 | 536 | 77 | ||
Set Membership: |
|