Search
Search

PPD5| Gene Symbol: PPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343192.1 | NP_001331121.1 | 2 - 3 | 308 | 69 | |
| NM_121183.5 | NP_196706.2 | 2 - 3 | 277 | 69 | ||
| GenBank mRNA | AY072412.1 | - | 2 - 3 | 270 | 69 | |
| AY114705.1 | - | 2 - 3 | 225 | 69 | ||
Set Membership: |
|