Search
Search

CID7| Gene Symbol: CID7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336049.1 | NP_001324897.1 | 3 - 4 | 1180 | 115 | |
| NM_001336050.1 | NP_001324896.1 | 3 - 4 | 1180 | 115 | ||
| NM_001336051.1 | NP_001324893.1 | 3 - 4 | 1120 | 115 | ||
| NM_001336052.1 | NP_001324892.1 | 3 - 4 | 1028 | 115 | ||
| NM_001336053.1 | NP_001324895.1 | 3 - 4 | 1207 | 115 | ||
| NM_001336054.1 | NP_001324894.1 | 3 - 4 | 1207 | 115 | ||
| NM_128185.3 | NP_180196.1 | 3 - 4 | 1039 | 115 | ||
| GenBank mRNA | BT030338.1 | - | 1 - 2 | 781 | 115 | |
Set Membership: |
|