Search
Search

TTN7| Gene Symbol: TTN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084499.2 | NP_001077968.1 | 20 - 21 | 2414 | 121 | |
| NM_001336108.1 | NP_001324367.1 | 20 - 21 | 2439 | 121 | ||
| NM_001336109.1 | NP_001324368.1 | 20 - 21 | 2373 | 121 | ||
| NM_128275.5 | NP_180285.4 | 20 - 21 | 2454 | 121 | ||
| GenBank mRNA | AK221283.1 | - | 20 - 21 | 2392 | 121 | |
Set Membership: |
|