Search
Search

TTN7| Gene Symbol: TTN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084499.2 | NP_001077968.1 | 25 - 26 | 3179 | 83 | |
| NM_001336108.1 | NP_001324367.1 | 25 - 26 | 3204 | 83 | ||
| NM_001336109.1 | NP_001324368.1 | 25 - 26 | 3138 | 83 | ||
| NM_128275.5 | NP_180285.4 | 25 - 26 | 3219 | 83 | ||
| GenBank mRNA | AK221283.1 | - | 25 - 26 | 3157 | 83 | |
Set Membership: |
|