Search
Search

LHW| Gene Symbol: LHW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036353.1 | NP_001031430.1 | 7 - 8 | 2126 | 99 | |
| NM_128281.2 | NP_565640.1 | 7 - 8 | 2089 | 99 | ||
| GenBank mRNA | AB493566.1 | - | 6 - 7 | 1677 | 99 | |
| AJ576043.1 | - | 6 - 7 | 1677 | 99 | ||
Set Membership: |
|