Search
Search

VLN1| Gene Symbol: VLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036367.2 | NP_001031444.2 | 7 - 8 | 919 | 92 | |
| NM_001202706.1 | NP_001189635.1 | 7 - 8 | 919 | 92 | ||
| NM_001336227.1 | NP_001324174.1 | 7 - 8 | 953 | 92 | ||
| NM_001336228.1 | NP_001324173.1 | 7 - 8 | 1004 | 92 | ||
| NM_128543.4 | NP_029567.1 | 7 - 8 | 938 | 92 | ||
Set Membership: |
|