Search
Search

HMT3| Gene Symbol: HMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113173.3 | NP_566715.1 | 3 - 4 | 759 | 137 | |
| GenBank mRNA | AF297394.1 | - | 3 - 4 | 464 | 137 | |
| AK118021.1 | - | 3 - 4 | 559 | 137 | ||
| BT005318.1 | - | 3 - 4 | 464 | 137 | ||
Set Membership: |
|