Search
Search

RNR2A| Gene Symbol: RNR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113261.4 | NP_189000.1 | 2 - 3 | 742 | 153 | |
| GenBank mRNA | AF372971.1 | - | 2 - 3 | 474 | 153 | |
| AY143893.1 | - | 2 - 3 | 319 | 153 | ||
Set Membership: |
|