Search
Search

TOPII| Gene Symbol: TOPII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125214.2 | NP_001118686.1 | 7 - 8 | 2019 | 133 | |
| NM_113294.3 | NP_189031.1 | 7 - 8 | 2029 | 133 | ||
| GenBank mRNA | L21015.1 | - | 7 - 8 | 1915 | 133 | |
Set Membership: |
|