Search
Search

MBF1C| Gene Symbol: MBF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113358.3 | NP_189093.1 | 1 | 879 | 102 | |
| GenBank mRNA | AK227957.1 | - | 1 | 467 | 102 | |
| BT004761.1 | - | 1 | 391 | 102 | ||
Set Membership: |
|