Search
Search

RIN4| Gene Symbol: RIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338741.1 | NP_001325873.1 | 2 - 3 | 1405 | 65 | |
| NM_113411.3 | NP_189143.2 | 3 - 4 | 1003 | 65 | ||
| GenBank mRNA | AK117488.1 | - | 3 - 4 | 674 | 65 | |
Set Membership: |
|