Search
Search

GLYR1| Gene Symbol: GLYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035688.1 | NP_001030765.1 | 1 - 2 | 184 | 112 | |
| NM_001338754.1 | NP_001326583.1 | 1 - 2 | 226 | 112 | ||
| NM_113449.4 | NP_566768.1 | 1 - 2 | 342 | 112 | ||
| GenBank mRNA | BT025039.1 | - | 1 - 2 | 105 | 112 | |
Set Membership: |
|