Search
Search

HMT-1| Gene Symbol: HMT-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203048.1 | NP_001189977.1 | 2 - 3 | 367 | 139 | |
| NM_113493.5 | NP_189219.1 | 2 - 3 | 446 | 139 | ||
| NM_202632.1 | NP_974361.1 | 2 - 3 | 353 | 139 | ||
| GenBank mRNA | AY065163.1 | - | 2 - 3 | 353 | 139 | |
| AY081604.1 | - | 2 - 3 | 228 | 139 | ||
Set Membership: |
|