Search
Search

RGS1| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338780.1 | NP_001327634.1 | 3 - 4 | 539 | 78 | |
| NM_113514.4 | NP_189238.2 | 3 - 4 | 539 | 78 | ||
| GenBank mRNA | AY150429.1 | - | 3 - 4 | 269 | 78 | |
Set Membership: |
|