Search
Search

CCB1| Gene Symbol: CCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113582.5 | NP_566797.1 | 3 - 4 | 942 | 155 | |
| GenBank mRNA | AK227251.1 | - | 3 - 4 | 797 | 155 | |
| AY087129.1 | - | 3 - 4 | 634 | 155 | ||
| AY140025.1 | - | 3 - 4 | 640 | 155 | ||
| BT006294.1 | - | 3 - 4 | 615 | 155 | ||
Set Membership: |
|