Search
Search

DXR| Gene Symbol: DXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125674.3 | NP_201085.1 | 3 - 4 | 551 | 75 | |
| GenBank mRNA | AF148852.1 | - | 3 - 4 | 459 | 75 | |
| AY045634.1 | - | 3 - 4 | 498 | 75 | ||
| AY050802.1 | - | 3 - 4 | 497 | 75 | ||
| AY054682.1 | - | 3 - 4 | 459 | 75 | ||
| AY081453.1 | - | 3 - 4 | 374 | 75 | ||
| AY084775.1 | - | 3 - 4 | 459 | 75 | ||
| AY091405.1 | - | 3 - 4 | 374 | 75 | ||
| AY098952.1 | - | 3 - 4 | 374 | 75 | ||
Set Membership: |
|