Search
Search

DXR| Gene Symbol: DXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203671.1 | NP_001190600.1 | 8 - 9 | 972 | 149 | |
| NM_125674.3 | NP_201085.1 | 7 - 8 | 957 | 149 | ||
| GenBank mRNA | AF148852.1 | - | 7 - 8 | 865 | 149 | |
| AY045634.1 | - | 7 - 8 | 904 | 149 | ||
| AY050802.1 | - | 7 - 8 | 903 | 149 | ||
| AY054682.1 | - | 7 - 8 | 865 | 149 | ||
| AY081453.1 | - | 7 - 8 | 780 | 149 | ||
| AY084775.1 | - | 7 - 8 | 865 | 149 | ||
| AY091405.1 | - | 7 - 8 | 780 | 149 | ||
| AY098952.1 | - | 7 - 8 | 780 | 149 | ||
Set Membership: |
|