Search
Search

FP3| Gene Symbol: FP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085321.2 | NP_001078790.1 | 3 - 4 | 701 | 89 | |
| NM_125748.3 | NP_568974.1 | 4 - 5 | 978 | 89 | ||
| GenBank mRNA | AF360327.1 | - | 4 - 5 | 625 | 89 | |
| AY056337.1 | - | 3 - 4 | 589 | 89 | ||
Set Membership: |
|