Search
Search

SHA1| Gene Symbol: SHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345611.1 | NP_001331890.1 | 2 - 3 | 1915 | 67 | |
| NM_125773.4 | NP_201183.2 | 2 - 3 | 1368 | 67 | ||
| GenBank mRNA | BT002521.1 | - | 2 - 3 | 1351 | 67 | |
| BT008456.1 | - | 1 - 2 | 910 | 67 | ||
Set Membership: |
|