Search
Search

CHR24| Gene Symbol: CHR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345616.1 | NP_001332133.1 | 12 - 13 | 2389 | 107 | |
| NM_125791.3 | NP_201200.2 | 12 - 13 | 2491 | 107 | ||
| GenBank mRNA | AF462829.1 | - | 12 - 13 | 2325 | 107 | |
| AY142010.1 | - | 12 - 13 | 2225 | 107 | ||
Set Membership: |
|