Search
Search

OPT4| Gene Symbol: OPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125837.4 | NP_201246.1 | 4 - 5 | 1926 | 89 | |
| GenBank mRNA | AK227021.1 | - | 4 - 5 | 1477 | 89 | |
| AY099843.1 | - | 4 - 5 | 1473 | 89 | ||
| BT000339.1 | - | 4 - 5 | 1399 | 89 | ||
Set Membership: |
|